| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016826.hg.1 |
ENST00000332591.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016826.hg.1 |
ENST00000375495.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016826.hg.1 |
ENST00000395505.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016826.hg.1 |
ENST00000395506.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016826.hg.1 |
NM_001007101.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016826.hg.1 |
NM_031486.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016826.hg.1 |
TC09001342.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000332591.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000332591.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000332591.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000332591.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000332591.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000332591.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000332591.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000375495.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000375495.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000375495.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000375495.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000375495.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000375495.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000375495.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000375495.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000395505.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000395505.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000395505.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000395505.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000395505.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000395505.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000395505.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000395505.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000395505.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001007101.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001007101.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001007101.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001007101.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001007101.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001007101.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001007101.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001007101.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261458.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261458.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261458.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261458.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261458.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261458.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261458.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261458.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261458.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261460.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261460.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261460.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261460.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261460.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261460.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261460.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261460.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261460.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_001261460.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_031486.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_031486.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_031486.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_031486.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_031486.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_031486.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_031486.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_031486.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010814.hg.1 |
NM_031486.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000332591.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000332591.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000332591.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000332591.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000332591.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000332591.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000332591.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000375495.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000375495.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000375495.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000375495.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000375495.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000375495.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000375495.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000375495.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000395505.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000395505.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000395505.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000395505.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000395505.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000395505.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000395505.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000395505.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000395505.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001007101.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001007101.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001007101.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001007101.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001007101.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001007101.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001007101.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001007101.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261458.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261458.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261458.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261458.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261458.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261458.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261458.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261458.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261458.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261460.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261460.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261460.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261460.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261460.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261460.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261460.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261460.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261460.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_001261460.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_031486.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_031486.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_031486.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_031486.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_031486.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_031486.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_031486.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_031486.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010814.hg.1 |
NM_031486.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173614.hg.1 |
ENST00000332591.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173614.hg.1 |
ENST00000375495.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173614.hg.1 |
ENST00000395505.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173614.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173614.hg.1 |
NM_001007101.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173614.hg.1 |
NM_001261458.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173614.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173614.hg.1 |
NM_001261460.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173614.hg.1 |
NM_031486.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173614.hg.1 |
ENST00000332591.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173614.hg.1 |
ENST00000375495.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173614.hg.1 |
ENST00000395505.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173614.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173614.hg.1 |
NM_001007101.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173614.hg.1 |
NM_001261458.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173614.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173614.hg.1 |
NM_001261460.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173614.hg.1 |
NM_031486.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173614.hg.1 |
TC0900010814.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900173618.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173618.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173618.hg.1 |
NM_001261460.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173618.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173618.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173618.hg.1 |
NM_001261460.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173618.hg.1 |
TC0900010814.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900173623.hg.1 |
ENST00000395505.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173623.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173623.hg.1 |
NM_001261458.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173623.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173623.hg.1 |
ENST00000395505.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173623.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173623.hg.1 |
NM_001261458.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173623.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173623.hg.1 |
TC0900010814.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900173616.hg.1 |
ENST00000332591.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173616.hg.1 |
ENST00000375495.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173616.hg.1 |
ENST00000395505.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173616.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173616.hg.1 |
NM_001007101.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173616.hg.1 |
NM_001261458.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173616.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173616.hg.1 |
NM_001261460.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173616.hg.1 |
NM_031486.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173616.hg.1 |
ENST00000332591.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173616.hg.1 |
ENST00000375495.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173616.hg.1 |
ENST00000395505.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173616.hg.1 |
ENST00000395506.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173616.hg.1 |
NM_001007101.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173616.hg.1 |
NM_001261458.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173616.hg.1 |
NM_001261459.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173616.hg.1 |
NM_001261460.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173616.hg.1 |
NM_031486.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173616.hg.1 |
TC0900010814.hg.1 |
Clariom D TC |