| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000367368.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000367368.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000367368.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000367368.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000463728.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000463728.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
ENST00000463728.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
ENST00000446954.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
ENST00000446954.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
ENST00000446954.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
ENST00000446954.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
ENST00000596229.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
ENST00000596229.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
ENST00000596229.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
ENST00000596229.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
ENST00000596229.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
ENST00000605899.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
ENST00000605899.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
ENST00000605899.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
ENST00000605899.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
ENST00000606915.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
ENST00000606915.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
ENST00000606915.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
ENST00000606915.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
ENST00000606915.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
ENST00000606915.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
ENST00000606915.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
NR_045126.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
NR_045126.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
NR_045126.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
NR_045126.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
NR_045126.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
NR_045126.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
NR_045126.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
NR_045127.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
NR_045127.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
NR_045127.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009813.hg.1 |
NR_045127.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027789.hg.1 |
ENST00000239367.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027789.hg.1 |
ENST00000367368.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027789.hg.1 |
ENST00000546019.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027789.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027789.hg.1 |
TC06002217.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027788.hg.1 |
ENST00000239367.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027788.hg.1 |
ENST00000367368.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027788.hg.1 |
ENST00000546019.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027788.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027788.hg.1 |
TC06002217.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027790.hg.1 |
ENST00000239367.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027790.hg.1 |
ENST00000367368.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027790.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027790.hg.1 |
TC06002217.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027791.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027791.hg.1 |
TC06002217.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600202855.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202855.hg.1 |
ENST00000367368.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202855.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600202855.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202855.hg.1 |
ENST00000367368.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202855.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600202855.hg.1 |
TC0600014357.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600202857.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600202857.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600202857.hg.1 |
TC0600014357.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
ENST00000446954.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
ENST00000446954.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
ENST00000446954.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
ENST00000446954.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
ENST00000596229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
ENST00000596229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
ENST00000596229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
ENST00000596229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
ENST00000596229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
ENST00000605899.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
ENST00000605899.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
ENST00000605899.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
ENST00000605899.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
ENST00000606915.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
ENST00000606915.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
ENST00000606915.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
ENST00000606915.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
ENST00000606915.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
ENST00000606915.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
ENST00000606915.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
NR_045126.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
NR_045126.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
NR_045126.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
NR_045126.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
NR_045126.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
NR_045126.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
NR_045126.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
NR_045127.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
NR_045127.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
NR_045127.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009813.hg.1 |
NR_045127.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000367368.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000367368.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000367368.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000367368.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000463728.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000463728.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
ENST00000463728.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014357.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600202848.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202848.hg.1 |
ENST00000367368.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202848.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600202848.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202848.hg.1 |
ENST00000367368.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202848.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600202848.hg.1 |
TC0600014357.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600202853.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202853.hg.1 |
ENST00000367368.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202853.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600202853.hg.1 |
ENST00000239367.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202853.hg.1 |
ENST00000367368.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202853.hg.1 |
NM_032832.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600202853.hg.1 |
TC0600014357.hg.1 |
Clariom D TC |