| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000610069.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000610069.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000610069.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
ENST00000610069.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009045.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22015001.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22015001.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22015001.hg.1 |
TC22000893.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22015000.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22015000.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22015000.hg.1 |
TC22000893.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2200165483.hg.1 |
TC2200009045.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200165483.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165483.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165483.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165483.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165483.hg.1 |
ENST00000610069.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165483.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165483.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165483.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165483.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165483.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165483.hg.1 |
ENST00000610069.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165483.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165482.hg.1 |
TC2200009045.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200165482.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165482.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165482.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165482.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165482.hg.1 |
ENST00000610069.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165482.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165482.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165482.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165482.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165482.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165482.hg.1 |
ENST00000610069.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165482.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165484.hg.1 |
TC2200009045.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200165484.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165484.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165484.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165484.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165484.hg.1 |
ENST00000610069.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165484.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165484.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165484.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165484.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165484.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165484.hg.1 |
ENST00000610069.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165484.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165485.hg.1 |
TC2200009045.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200165485.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165485.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165485.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165485.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165485.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165485.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000380990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000609786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000610069.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000610069.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000610069.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
ENST00000610069.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
NR_033377.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009045.hg.1 |
NR_033377.1 |
Refseq |