| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01034545.hg.1 |
ENST00000335496.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01034545.hg.1 |
ENST00000466212.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01034545.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01034545.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01034545.hg.1 |
TC01002263.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000491418.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000491418.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000491418.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000491418.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000612240.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000612240.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000612240.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000612240.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000612240.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100196148.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100196148.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100196148.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100196148.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100196148.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100196148.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100196148.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100196148.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100196148.hg.1 |
TC0100013042.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000491418.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000491418.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000491418.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000491418.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000612240.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000612240.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000612240.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000612240.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000612240.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |