| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000592227.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000592227.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000592227.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000592227.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
ENST00000592227.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19003550.hg.1 |
ENST00000254323.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19003550.hg.1 |
ENST00000440752.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19003550.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19003550.hg.1 |
TC19000241.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19003551.hg.1 |
ENST00000254323.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19003551.hg.1 |
ENST00000440752.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19003551.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19003551.hg.1 |
TC19000241.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900151457.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900151457.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900151457.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900151457.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900151457.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900151457.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900151457.hg.1 |
TC1900007142.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000592227.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000592227.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000592227.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000592227.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
ENST00000592227.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007142.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900151455.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900151455.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900151455.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900151455.hg.1 |
ENST00000254323.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900151455.hg.1 |
ENST00000590508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900151455.hg.1 |
NM_023072.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900151455.hg.1 |
TC1900007142.hg.1 |
Clariom D TC |