| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01040613.hg.1 |
TC01002608.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01040613.hg.1 |
ENST00000341771.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01040613.hg.1 |
ENST00000372083.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01040613.hg.1 |
ENST00000372084.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01040613.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01040613.hg.1 |
ENST00000486676.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01040613.hg.1 |
ENST00000538496.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01040613.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000372084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000372084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000372084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000372084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000372084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000372084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000372084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000372084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000372084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000372084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000372084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000493974.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000493974.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
ENST00000493974.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000372084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000372084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000372084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000372084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000372084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000372084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000372084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000372084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000372084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000372084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000372084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000493974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000493974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
ENST00000493974.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014009.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100204755.hg.1 |
ENST00000372084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100204755.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100204755.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100204755.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100204755.hg.1 |
ENST00000372084.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100204755.hg.1 |
ENST00000372086.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100204755.hg.1 |
ENST00000486676.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100204755.hg.1 |
NM_007170.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100204755.hg.1 |
TC0100014009.hg.1 |
Clariom D TC |