| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19019640.hg.1 |
ENST00000004982.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19019640.hg.1 |
ENST00000416180.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19019640.hg.1 |
NM_144617.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19019640.hg.1 |
TC19001453.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19019641.hg.1 |
ENST00000004982.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19019641.hg.1 |
NM_144617.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19019641.hg.1 |
TC19001453.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19019642.hg.1 |
ENST00000004982.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19019642.hg.1 |
ENST00000416180.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19019642.hg.1 |
NM_144617.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19019642.hg.1 |
TC19001453.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900185908.hg.1 |
ENST00000004982.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900185908.hg.1 |
ENST00000592984.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900185908.hg.1 |
NM_144617.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900185908.hg.1 |
ENST00000004982.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900185908.hg.1 |
ENST00000592984.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900185908.hg.1 |
NM_144617.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900185908.hg.1 |
TC1900010494.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900185907.hg.1 |
ENST00000004982.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900185907.hg.1 |
ENST00000592984.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900185907.hg.1 |
NM_144617.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900185907.hg.1 |
ENST00000004982.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900185907.hg.1 |
ENST00000592984.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900185907.hg.1 |
NM_144617.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900185907.hg.1 |
TC1900010494.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900185909.hg.1 |
ENST00000004982.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900185909.hg.1 |
ENST00000587965.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900185909.hg.1 |
ENST00000592984.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900185909.hg.1 |
NM_144617.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900185909.hg.1 |
ENST00000004982.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900185909.hg.1 |
ENST00000587965.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900185909.hg.1 |
ENST00000592984.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900185909.hg.1 |
NM_144617.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900185909.hg.1 |
TC1900010494.hg.1 |
Clariom D TC |