| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010049.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010049.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010049.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010049.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010049.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010049.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010049.hg.1 |
ENST00000556881.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010049.hg.1 |
ENST00000556881.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010049.hg.1 |
ENST00000556881.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010049.hg.1 |
ENST00000556881.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010049.hg.1 |
ENST00000556881.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010049.hg.1 |
NM_001304461.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010049.hg.1 |
NM_001304461.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010049.hg.1 |
NM_001304461.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010049.hg.1 |
NM_001304461.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010049.hg.1 |
NM_001304461.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010049.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010049.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010049.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010049.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010049.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010049.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14016570.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14016570.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14016570.hg.1 |
TC14001469.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14016564.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14016564.hg.1 |
ENST00000556881.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14016564.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14016564.hg.1 |
TC14001469.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14016569.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14016569.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14016569.hg.1 |
TC14001469.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14016565.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14016565.hg.1 |
ENST00000556881.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14016565.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14016565.hg.1 |
TC14001469.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14016566.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14016566.hg.1 |
ENST00000556881.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14016566.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14016566.hg.1 |
TC14001469.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1400174469.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400174469.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400174469.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400174469.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400174469.hg.1 |
TC1400010049.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400174470.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400174470.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400174470.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400174470.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400174470.hg.1 |
TC1400010049.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400174465.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400174465.hg.1 |
ENST00000556881.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400174465.hg.1 |
NM_001304461.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400174465.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400174465.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400174465.hg.1 |
ENST00000556881.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400174465.hg.1 |
NM_001304461.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400174465.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400174465.hg.1 |
TC1400010049.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1400010049.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010049.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010049.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010049.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010049.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010049.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010049.hg.1 |
ENST00000556881.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010049.hg.1 |
ENST00000556881.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010049.hg.1 |
ENST00000556881.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010049.hg.1 |
ENST00000556881.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010049.hg.1 |
ENST00000556881.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010049.hg.1 |
NM_001304461.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010049.hg.1 |
NM_001304461.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010049.hg.1 |
NM_001304461.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010049.hg.1 |
NM_001304461.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010049.hg.1 |
NM_001304461.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010049.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010049.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010049.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010049.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010049.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010049.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400174463.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400174463.hg.1 |
ENST00000556881.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400174463.hg.1 |
NM_001304461.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400174463.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400174463.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400174463.hg.1 |
ENST00000556881.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400174463.hg.1 |
NM_001304461.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400174463.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400174463.hg.1 |
TC1400010049.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400174464.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400174464.hg.1 |
ENST00000556881.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400174464.hg.1 |
NM_001304461.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400174464.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400174464.hg.1 |
ENST00000341228.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400174464.hg.1 |
ENST00000556881.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400174464.hg.1 |
NM_001304461.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400174464.hg.1 |
NM_173850.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400174464.hg.1 |
TC1400010049.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400174468.hg.1 |
ENST00000556881.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400174468.hg.1 |
NM_001304461.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400174468.hg.1 |
ENST00000556881.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400174468.hg.1 |
NM_001304461.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400174468.hg.1 |
TC1400010049.hg.1 |
Clariom D TC |