| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01057518.hg.1 |
ENST00000367145.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01057518.hg.1 |
NM_033102.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01057518.hg.1 |
TC01003750.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017073.hg.1 |
ENST00000367145.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017073.hg.1 |
ENST00000367145.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017073.hg.1 |
ENST00000367145.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017073.hg.1 |
ENST00000367145.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017073.hg.1 |
ENST00000367145.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017073.hg.1 |
ENST00000367145.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017073.hg.1 |
ENST00000367145.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017073.hg.1 |
ENST00000460934.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017073.hg.1 |
ENST00000460934.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017073.hg.1 |
ENST00000460934.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017073.hg.1 |
ENST00000460934.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017073.hg.1 |
NM_033102.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017073.hg.1 |
NM_033102.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017073.hg.1 |
NM_033102.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017073.hg.1 |
NM_033102.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017073.hg.1 |
NM_033102.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017073.hg.1 |
NM_033102.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017073.hg.1 |
NM_033102.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017073.hg.1 |
NM_033102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017073.hg.1 |
ENST00000367145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017073.hg.1 |
ENST00000367145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017073.hg.1 |
ENST00000367145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017073.hg.1 |
ENST00000367145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017073.hg.1 |
ENST00000367145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017073.hg.1 |
ENST00000367145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017073.hg.1 |
ENST00000367145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017073.hg.1 |
ENST00000460934.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017073.hg.1 |
ENST00000460934.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017073.hg.1 |
ENST00000460934.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017073.hg.1 |
ENST00000460934.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017073.hg.1 |
NM_033102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017073.hg.1 |
NM_033102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017073.hg.1 |
NM_033102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017073.hg.1 |
NM_033102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017073.hg.1 |
NM_033102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017073.hg.1 |
NM_033102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017073.hg.1 |
NM_033102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017073.hg.1 |
NM_033102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100229367.hg.1 |
ENST00000367145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100229367.hg.1 |
NM_033102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100229367.hg.1 |
ENST00000367145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100229367.hg.1 |
NM_033102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100229367.hg.1 |
TC0100017073.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100229366.hg.1 |
ENST00000367145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100229366.hg.1 |
NM_033102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100229366.hg.1 |
ENST00000367145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100229366.hg.1 |
NM_033102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100229366.hg.1 |
TC0100017073.hg.1 |
Clariom D TC |