| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000528184.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000528184.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000528184.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000528184.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000528184.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000528511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000528511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000528511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000528511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000528511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000528511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000528511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000528511.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000528560.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000528560.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000528560.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000528560.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000529513.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000529513.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000529513.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000529513.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000529513.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000529513.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000529513.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000529513.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000529513.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000529513.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000531488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000531488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000531488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000532852.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000532852.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000532852.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000532852.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000532852.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000532852.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000533047.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000533047.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000533047.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000533047.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000533047.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000533047.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000533047.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000534704.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000534704.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000534704.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000534704.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000534704.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000534704.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
ENST00000534704.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011535.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11024427.hg.1 |
ENST00000528511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11024427.hg.1 |
ENST00000529513.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11024427.hg.1 |
ENST00000529844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11024427.hg.1 |
ENST00000530333.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11024427.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11024427.hg.1 |
TC11002048.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1100188726.hg.1 |
ENST00000528184.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100188726.hg.1 |
ENST00000528511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100188726.hg.1 |
ENST00000529513.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100188726.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100188726.hg.1 |
ENST00000532852.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100188726.hg.1 |
ENST00000533047.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100188726.hg.1 |
ENST00000534704.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100188726.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100188726.hg.1 |
ENST00000528184.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100188726.hg.1 |
ENST00000528511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100188726.hg.1 |
ENST00000529513.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100188726.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100188726.hg.1 |
ENST00000532852.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100188726.hg.1 |
ENST00000533047.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100188726.hg.1 |
ENST00000534704.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100188726.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100188726.hg.1 |
TC1100011535.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000528184.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000528184.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000528184.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000528184.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000528184.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000528511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000528511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000528511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000528511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000528511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000528511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000528511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000528511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000528560.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000528560.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000528560.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000528560.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000529513.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000529513.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000529513.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000529513.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000529513.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000529513.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000529513.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000529513.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000529513.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000529513.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000531488.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000531488.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000531488.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000532852.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000532852.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000532852.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000532852.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000532852.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000532852.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000533047.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000533047.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000533047.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000533047.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000533047.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000533047.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000533047.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000534704.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000534704.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000534704.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000534704.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000534704.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000534704.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
ENST00000534704.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011535.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100188727.hg.1 |
ENST00000528184.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100188727.hg.1 |
ENST00000528511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100188727.hg.1 |
ENST00000529513.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100188727.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100188727.hg.1 |
ENST00000533047.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100188727.hg.1 |
ENST00000534704.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100188727.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100188727.hg.1 |
ENST00000528184.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100188727.hg.1 |
ENST00000528511.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100188727.hg.1 |
ENST00000529513.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100188727.hg.1 |
ENST00000529844.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100188727.hg.1 |
ENST00000533047.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100188727.hg.1 |
ENST00000534704.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100188727.hg.1 |
NR_029192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100188727.hg.1 |
TC1100011535.hg.1 |
Clariom D TC |