| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19007908.hg.1 |
ENST00000331105.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19007908.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19007908.hg.1 |
TC19000573.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000439903.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000439903.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000439903.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000439903.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000439903.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000439903.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000526093.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000526093.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000526093.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000526093.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000526093.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000526093.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000532164.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000532164.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000532164.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000532164.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000532164.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000532164.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000532164.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000532164.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000532164.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000532164.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000532164.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000532164.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000534009.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000534009.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000534009.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000534009.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000534009.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
ENST00000534009.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000439903.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000439903.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000439903.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000439903.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000439903.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000439903.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000526093.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000526093.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000526093.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000526093.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000526093.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000526093.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000532164.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000532164.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000532164.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000532164.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000532164.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000532164.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000532164.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000532164.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000532164.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000532164.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000532164.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000532164.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000534009.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000534009.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000534009.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000534009.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000534009.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
ENST00000534009.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011738.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900200963.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900200963.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900200963.hg.1 |
ENST00000532164.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900200963.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900200963.hg.1 |
ENST00000331105.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900200963.hg.1 |
ENST00000531409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900200963.hg.1 |
ENST00000532164.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900200963.hg.1 |
NM_000774.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900200963.hg.1 |
TC1900011738.hg.1 |
Clariom D TC |