| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01048234.hg.1 |
ENST00000235933.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01048234.hg.1 |
ENST00000369288.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01048234.hg.1 |
ENST00000369290.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01048234.hg.1 |
ENST00000401557.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01048234.hg.1 |
NM_007053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01048234.hg.1 |
TC01003113.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000235933.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000235933.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000235933.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000235933.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000235933.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000235933.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000369288.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000369288.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000369288.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000369288.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000369288.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000369288.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000369288.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000401557.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000401557.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000401557.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000401557.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000584442.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000584442.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000616463.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000616463.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000616463.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000616463.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000616463.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000616463.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000616463.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
ENST00000616463.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
NM_007053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
NM_007053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
NM_007053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
NM_007053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
NM_007053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
NM_007053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
NM_007053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
NM_007053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
NM_007053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
NM_007053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
NR_103845.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
NR_103845.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
NR_103845.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
NR_103845.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
NR_103845.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
NR_103845.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009742.hg.1 |
NR_103845.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000235933.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000235933.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000235933.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000235933.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000235933.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000235933.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000369288.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000369288.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000369288.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000369288.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000369288.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000369288.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000369288.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000401557.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000401557.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000401557.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000401557.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000584442.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000584442.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000616463.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000616463.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000616463.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000616463.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000616463.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000616463.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000616463.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
ENST00000616463.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
NM_007053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
NM_007053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
NM_007053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
NM_007053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
NM_007053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
NM_007053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
NM_007053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
NM_007053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
NM_007053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
NM_007053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
NR_103845.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
NR_103845.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
NR_103845.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
NR_103845.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
NR_103845.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
NR_103845.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009742.hg.1 |
NR_103845.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100170049.hg.1 |
ENST00000235933.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100170049.hg.1 |
ENST00000369288.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100170049.hg.1 |
ENST00000401557.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100170049.hg.1 |
ENST00000584442.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100170049.hg.1 |
ENST00000616463.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100170049.hg.1 |
NM_007053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100170049.hg.1 |
NR_103845.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100170049.hg.1 |
ENST00000235933.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100170049.hg.1 |
ENST00000369288.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100170049.hg.1 |
ENST00000401557.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100170049.hg.1 |
ENST00000584442.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100170049.hg.1 |
ENST00000616463.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100170049.hg.1 |
NM_007053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100170049.hg.1 |
NR_103845.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100170049.hg.1 |
TC0100009742.hg.1 |
Clariom D TC |