| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000255616.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000255616.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000255616.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000255616.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000255616.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000255616.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000255616.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000255616.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000255616.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000255616.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000593661.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000593661.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000593661.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000593661.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000593661.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000593661.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000593661.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000594748.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000594748.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000594748.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000594748.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000594748.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000594748.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000595348.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000595348.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000595348.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000595348.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000595348.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000596772.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000596772.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000596772.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000596772.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000596772.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000596772.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000596772.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000599379.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000599379.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000599379.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000599379.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000600906.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000600906.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000600906.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000602128.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000602128.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000602128.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000602128.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000602128.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
ENST00000602128.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
NM_032370.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
NM_032370.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
NM_032370.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
NM_032370.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
NM_032370.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
NM_032370.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
NM_032370.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
NM_032370.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
NM_032370.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009547.hg.1 |
NM_032370.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015581.hg.1 |
ENST00000255616.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015581.hg.1 |
ENST00000393927.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015581.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015581.hg.1 |
NM_032370.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19015581.hg.1 |
TC19001134.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900176749.hg.1 |
ENST00000255616.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176749.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176749.hg.1 |
ENST00000599379.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176749.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900176749.hg.1 |
NM_032370.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900176749.hg.1 |
TC1900009547.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900176749.hg.1 |
ENST00000255616.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176749.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176749.hg.1 |
ENST00000599379.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900176749.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900176749.hg.1 |
NM_032370.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000255616.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000255616.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000255616.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000255616.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000255616.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000255616.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000255616.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000255616.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000255616.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000255616.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000393927.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000593661.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000593661.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000593661.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000593661.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000593661.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000593661.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000593661.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000594748.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000594748.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000594748.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000594748.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000594748.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000594748.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000595348.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000595348.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000595348.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000595348.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000595348.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000596772.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000596772.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000596772.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000596772.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000596772.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000596772.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000596772.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000599379.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000599379.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000599379.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000599379.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000600906.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000600906.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000600906.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000602128.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000602128.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000602128.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000602128.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000602128.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
ENST00000602128.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
NM_001146175.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
NM_032370.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
NM_032370.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
NM_032370.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
NM_032370.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
NM_032370.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
NM_032370.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
NM_032370.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
NM_032370.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
NM_032370.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009547.hg.1 |
NM_032370.2 |
Refseq |