| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000513296.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000513296.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000513296.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000513296.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000513296.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000519933.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000519933.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000519933.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000519933.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000519933.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000523688.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000523688.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000523688.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000523688.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
ENST00000523688.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05000299.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05000299.hg.1 |
ENST00000296849.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05000299.hg.1 |
ENST00000537972.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05000299.hg.1 |
NM_033120.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05000299.hg.1 |
TC05000015.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000513296.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000513296.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000513296.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000513296.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000513296.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000519933.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000519933.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000519933.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000519933.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000519933.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000523688.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000523688.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000523688.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000523688.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
ENST00000523688.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006479.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500144561.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500144561.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500144561.hg.1 |
ENST00000519933.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500144561.hg.1 |
ENST00000523688.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500144561.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500144561.hg.1 |
ENST00000274150.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500144561.hg.1 |
ENST00000296849.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500144561.hg.1 |
ENST00000519933.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500144561.hg.1 |
ENST00000523688.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500144561.hg.1 |
NM_001271082.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500144561.hg.1 |
TC0500006479.hg.1 |
Clariom D TC |