Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000228682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000228682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000228682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000228682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000228682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000228682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000228682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000228682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000228682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000228682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000228682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000228682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000228682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000228682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000228682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000228682.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000527742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000527742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000527742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000527742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000528432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000528432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000528432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000528432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000528432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000528432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000528432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000528467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000528467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000528467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000528467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000528467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000528467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000528467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000528467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000528467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000528467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000528467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000530789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000530789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000530789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000530789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000530789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000532291.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000532291.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000532291.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000532291.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000532291.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000532291.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000532291.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000532291.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000543426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000543426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000543426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000543426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000543426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000543426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000543426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000543426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000543426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000543426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000543426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000543426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000543426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000546141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000546141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000546141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000546141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000546141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000546141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000546141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000546141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000546141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000546141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000546141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000546141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000546141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
ENST00000546141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001160045.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001160045.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001160045.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001160045.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001160045.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001160045.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001160045.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001160045.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001160045.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001160045.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001160045.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001160045.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001160045.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001167609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001167609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001167609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001167609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001167609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001167609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001167609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001167609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001167609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001167609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001167609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001167609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001167609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_001167609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_005269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_005269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_005269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_005269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_005269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_005269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_005269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_005269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_005269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_005269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_005269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_005269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_005269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_005269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_005269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007882.hg.1 |
NM_005269.2 |
Refseq |