| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011917.hg.1 |
ENST00000434401.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011917.hg.1 |
NR_038341.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011917.hg.1 |
TC20000872.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011916.hg.1 |
ENST00000434401.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011916.hg.1 |
NR_038341.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011916.hg.1 |
TC20000872.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011918.hg.1 |
NR_038341.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011918.hg.1 |
TC20000872.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009205.hg.1 |
ENST00000434401.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009205.hg.1 |
ENST00000434401.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009205.hg.1 |
ENST00000434401.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009205.hg.1 |
ENST00000434401.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009205.hg.1 |
ENST00000434401.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009205.hg.1 |
ENST00000434401.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009205.hg.1 |
ENST00000445571.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009205.hg.1 |
ENST00000445571.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009205.hg.1 |
ENST00000445571.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009205.hg.1 |
NR_038341.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009205.hg.1 |
NR_038341.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009205.hg.1 |
NR_038341.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009205.hg.1 |
NR_038341.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009205.hg.1 |
NR_038341.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009205.hg.1 |
NR_038341.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009205.hg.1 |
NR_038341.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009205.hg.1 |
ENST00000434401.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009205.hg.1 |
ENST00000434401.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009205.hg.1 |
ENST00000434401.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009205.hg.1 |
ENST00000434401.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009205.hg.1 |
ENST00000434401.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009205.hg.1 |
ENST00000434401.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009205.hg.1 |
ENST00000445571.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009205.hg.1 |
ENST00000445571.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009205.hg.1 |
ENST00000445571.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009205.hg.1 |
NR_038341.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009205.hg.1 |
NR_038341.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009205.hg.1 |
NR_038341.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009205.hg.1 |
NR_038341.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009205.hg.1 |
NR_038341.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009205.hg.1 |
NR_038341.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009205.hg.1 |
NR_038341.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000164406.hg.1 |
ENST00000434401.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164406.hg.1 |
NR_038341.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000164406.hg.1 |
TC2000009205.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000164406.hg.1 |
ENST00000434401.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164406.hg.1 |
NR_038341.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000164407.hg.1 |
ENST00000434401.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164407.hg.1 |
NR_038341.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000164407.hg.1 |
TC2000009205.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000164407.hg.1 |
ENST00000434401.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164407.hg.1 |
NR_038341.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000164408.hg.1 |
NR_038341.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000164408.hg.1 |
TC2000009205.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000164408.hg.1 |
NR_038341.1 |
Refseq |