| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010448.hg.1 |
ENST00000414116.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010448.hg.1 |
ENST00000414116.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010448.hg.1 |
ENST00000414116.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010448.hg.1 |
ENST00000414116.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010448.hg.1 |
ENST00000414116.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010448.hg.1 |
ENST00000414116.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010448.hg.1 |
ENST00000414116.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010448.hg.1 |
NR_122075.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010448.hg.1 |
NR_122075.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010448.hg.1 |
NR_122075.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010448.hg.1 |
NR_122075.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010448.hg.1 |
NR_122075.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010448.hg.1 |
NR_122075.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010448.hg.1 |
NR_122075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010448.hg.1 |
ENST00000414116.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010448.hg.1 |
ENST00000414116.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010448.hg.1 |
ENST00000414116.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010448.hg.1 |
ENST00000414116.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010448.hg.1 |
ENST00000414116.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010448.hg.1 |
ENST00000414116.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010448.hg.1 |
ENST00000414116.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010448.hg.1 |
NR_122075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010448.hg.1 |
NR_122075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010448.hg.1 |
NR_122075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010448.hg.1 |
NR_122075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010448.hg.1 |
NR_122075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010448.hg.1 |
NR_122075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010448.hg.1 |
NR_122075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174324.hg.1 |
ENST00000414116.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174324.hg.1 |
NR_122075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174324.hg.1 |
ENST00000414116.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174324.hg.1 |
NR_122075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174324.hg.1 |
TC0700010448.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700174327.hg.1 |
ENST00000414116.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174327.hg.1 |
NR_122075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174327.hg.1 |
ENST00000414116.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174327.hg.1 |
NR_122075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174327.hg.1 |
TC0700010448.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700174325.hg.1 |
ENST00000414116.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174325.hg.1 |
NR_122075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174325.hg.1 |
ENST00000414116.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174325.hg.1 |
NR_122075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174325.hg.1 |
TC0700010448.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700174326.hg.1 |
ENST00000414116.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174326.hg.1 |
NR_122075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174326.hg.1 |
ENST00000414116.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174326.hg.1 |
NR_122075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174326.hg.1 |
TC0700010448.hg.1 |
Clariom D TC |