| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
ENST00000417315.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
ENST00000417315.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
ENST00000433637.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
ENST00000433637.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
ENST00000433637.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
ENST00000433637.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
ENST00000457513.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
ENST00000457513.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
ENST00000457513.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
ENST00000574739.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
ENST00000574739.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
ENST00000574739.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
ENST00000574739.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
ENST00000574739.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600161467.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600161467.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600161467.hg.1 |
TC0600008884.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
ENST00000417315.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
ENST00000417315.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
ENST00000433637.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
ENST00000433637.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
ENST00000433637.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
ENST00000433637.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
ENST00000457513.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
ENST00000457513.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
ENST00000457513.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
ENST00000574739.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
ENST00000574739.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
ENST00000574739.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
ENST00000574739.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
ENST00000574739.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008884.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600161465.hg.1 |
ENST00000417315.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600161465.hg.1 |
ENST00000433637.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600161465.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600161465.hg.1 |
ENST00000417315.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600161465.hg.1 |
ENST00000433637.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600161465.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600161465.hg.1 |
TC0600008884.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600161469.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600161469.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600161469.hg.1 |
TC0600008884.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600161464.hg.1 |
ENST00000433637.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600161464.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600161464.hg.1 |
ENST00000433637.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600161464.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600161464.hg.1 |
TC0600008884.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600161466.hg.1 |
ENST00000433637.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600161466.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600161466.hg.1 |
ENST00000433637.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600161466.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600161466.hg.1 |
TC0600008884.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600161468.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600161468.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600161468.hg.1 |
TC0600008884.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600161470.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600161470.hg.1 |
NR_110757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600161470.hg.1 |
TC0600008884.hg.1 |
Clariom D TC |