| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009283.hg.1 |
ENST00000430381.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009283.hg.1 |
ENST00000430381.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009283.hg.1 |
ENST00000430381.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009283.hg.1 |
ENST00000430381.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009283.hg.1 |
ENST00000430381.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006467.hg.1 |
ENST00000382988.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006467.hg.1 |
ENST00000382988.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006467.hg.1 |
NR_126054.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006467.hg.1 |
NR_126054.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006467.hg.1 |
NR_126054.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006467.hg.1 |
NR_126054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300144173.hg.1 |
ENST00000382988.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144173.hg.1 |
NR_126054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300144173.hg.1 |
ENST00000382988.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144173.hg.1 |
NR_126054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300144173.hg.1 |
TC1300006467.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300144174.hg.1 |
NR_126054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300144174.hg.1 |
NR_126054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300144174.hg.1 |
TC1300006467.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1300006467.hg.1 |
ENST00000382988.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006467.hg.1 |
ENST00000382988.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006467.hg.1 |
NR_126054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006467.hg.1 |
NR_126054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006467.hg.1 |
NR_126054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006467.hg.1 |
NR_126054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200009283.hg.1 |
ENST00000430381.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009283.hg.1 |
ENST00000430381.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009283.hg.1 |
ENST00000430381.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009283.hg.1 |
ENST00000430381.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009283.hg.1 |
ENST00000430381.1 |
Ensembl |