| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12002159.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12002159.hg.1 |
ENST00000541162.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12002159.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12002159.hg.1 |
TC12000170.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12002158.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12002158.hg.1 |
ENST00000541162.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12002158.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12002158.hg.1 |
TC12000170.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000541162.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000541162.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000541162.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000541162.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000545239.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000545239.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000545239.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
ENST00000545239.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200147964.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147964.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147964.hg.1 |
ENST00000545239.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147964.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200147964.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147964.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147964.hg.1 |
ENST00000545239.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147964.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200147964.hg.1 |
TC1200006849.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200147965.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147965.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147965.hg.1 |
ENST00000541162.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147965.hg.1 |
ENST00000545239.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147965.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200147965.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147965.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147965.hg.1 |
ENST00000541162.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147965.hg.1 |
ENST00000545239.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147965.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200147965.hg.1 |
TC1200006849.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000541162.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000541162.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000541162.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000541162.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000545239.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000545239.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000545239.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
ENST00000545239.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006849.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200147966.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147966.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147966.hg.1 |
ENST00000541162.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147966.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200147966.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147966.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147966.hg.1 |
ENST00000541162.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147966.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200147966.hg.1 |
TC1200006849.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200147967.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147967.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147967.hg.1 |
ENST00000541162.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147967.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200147967.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147967.hg.1 |
ENST00000499291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147967.hg.1 |
ENST00000541162.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147967.hg.1 |
NR_033890.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200147967.hg.1 |
TC1200006849.hg.1 |
Clariom D TC |