| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009367.hg.1 |
ENST00000455438.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009367.hg.1 |
ENST00000455438.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009367.hg.1 |
ENST00000455438.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009367.hg.1 |
ENST00000455438.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009367.hg.1 |
ENST00000455438.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009367.hg.1 |
ENST00000455438.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009367.hg.1 |
ENST00000455438.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009367.hg.1 |
ENST00000455438.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009367.hg.1 |
NR_110412.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009367.hg.1 |
NR_110412.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009367.hg.1 |
NR_110412.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009367.hg.1 |
NR_110412.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009367.hg.1 |
NR_110412.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009367.hg.1 |
NR_110412.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009367.hg.1 |
NR_110412.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009367.hg.1 |
NR_110412.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00164201.hg.1 |
ENST00000455438.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00164201.hg.1 |
NR_110412.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00164201.hg.1 |
ENST00000455438.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00164201.hg.1 |
NR_110412.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00164201.hg.1 |
TC0X00009367.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00164203.hg.1 |
ENST00000455438.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00164203.hg.1 |
NR_110412.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00164203.hg.1 |
ENST00000455438.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00164203.hg.1 |
NR_110412.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00164203.hg.1 |
TC0X00009367.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00164200.hg.1 |
ENST00000455438.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00164200.hg.1 |
NR_110412.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00164200.hg.1 |
ENST00000455438.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00164200.hg.1 |
NR_110412.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00164200.hg.1 |
TC0X00009367.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00009367.hg.1 |
ENST00000455438.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009367.hg.1 |
ENST00000455438.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009367.hg.1 |
ENST00000455438.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009367.hg.1 |
ENST00000455438.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009367.hg.1 |
ENST00000455438.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009367.hg.1 |
ENST00000455438.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009367.hg.1 |
ENST00000455438.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009367.hg.1 |
ENST00000455438.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009367.hg.1 |
NR_110412.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009367.hg.1 |
NR_110412.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009367.hg.1 |
NR_110412.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009367.hg.1 |
NR_110412.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009367.hg.1 |
NR_110412.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009367.hg.1 |
NR_110412.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009367.hg.1 |
NR_110412.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009367.hg.1 |
NR_110412.1 |
Refseq |