| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07029597.hg.1 |
TC07001907.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07029597.hg.1 |
ENST00000242365.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07029597.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07029597.hg.1 |
ENST00000440172.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07029597.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07029597.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012755.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700191962.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700191962.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700191962.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700191962.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700191962.hg.1 |
ENST00000422774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700191962.hg.1 |
ENST00000440172.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700191962.hg.1 |
NM_001164665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700191962.hg.1 |
NM_020910.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700191962.hg.1 |
TC0700012755.hg.1 |
Clariom D TC |