Search
Search

MTARC2| Gene Symbol: MTARC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317338.2 | NP_001304267.1 | 3 - 4 | 820 | 60 | |
| NM_001331042.2 | NP_001317971.1 | 3 - 4 | 820 | 60 | ||
| NM_017898.5 | NP_060368.2 | 3 - 4 | 820 | 60 | ||
| XM_011509684.2 | XP_011507986.1 | 3 - 4 | 420 | 60 | ||
| XM_047423617.1 | XP_047279573.1 | 2 - 3 | 260 | 60 | ||
| GenBank mRNA | AK000612.1 | - | 1 - 2 | 35 | 60 | |
| AL136931.1 | - | 3 - 4 | 873 | 60 | ||
| BC010366.1 | - | 2 - 3 | 230 | 60 | ||
| BC011973.1 | - | 3 - 4 | 808 | 60 | ||
| BC015829.2 | - | 1 - 2 | 166 | 60 | ||
| BC016859.1 | - | 3 - 4 | 798 | 60 | ||
| BP194343.1 | - | 3 - 4 | 502 | 60 | ||
Set Membership: |
|