| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000423741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000423741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000423741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000423741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000423741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000423741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000423741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000423741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000423741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000451101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000451101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000451101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000451101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000451101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000537457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000537457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000537457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000537457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000537457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000537457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000537457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
ENST00000537457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200144879.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200144879.hg.1 |
ENST00000451101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200144879.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200144879.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200144879.hg.1 |
ENST00000451101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200144879.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200144879.hg.1 |
TC0200006540.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200144872.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200144872.hg.1 |
ENST00000537457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200144872.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200144872.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200144872.hg.1 |
ENST00000537457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200144872.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200144872.hg.1 |
TC0200006540.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200144870.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200144870.hg.1 |
ENST00000423741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200144870.hg.1 |
ENST00000537457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200144870.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200144870.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200144870.hg.1 |
ENST00000423741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200144870.hg.1 |
ENST00000537457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200144870.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200144870.hg.1 |
TC0200006540.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200144878.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200144878.hg.1 |
ENST00000451101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200144878.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200144878.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200144878.hg.1 |
ENST00000451101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200144878.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200144878.hg.1 |
TC0200006540.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000399143.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000423741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000423741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000423741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000423741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000423741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000423741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000423741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000423741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000423741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000451101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000451101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000451101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000451101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000451101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000537457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000537457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000537457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000537457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000537457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000537457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000537457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
ENST00000537457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006540.hg.1 |
NM_001287444.1 |
Refseq |