| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16734048 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513985 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513984 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513978 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513958 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513939 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513988 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513974 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513980 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513953 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29976037 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16734059 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16734056 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513965 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29976016 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513966 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513931 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513976 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513977 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513962 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513979 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16734034 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513971 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513934 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513947 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513936 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513950 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513959 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29976064 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513946 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513981 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29976053 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513940 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513990 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16734060 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513945 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513944 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16734026 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16734028 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513938 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29976073 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16734039 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29976068 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513972 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513943 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513963 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513948 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513956 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513975 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16734018 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16734055 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16734051 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513970 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29976050 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513961 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513987 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09513935 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29976081 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
114119 |
XM_936738.2 |
refseq_mrna_human_XM |
| Agilent SurePrint G3 Human CGH Microarray 8x60K |
6811 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
934288 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
695226 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
22211 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
266507 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P106258 |
- |
- |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_23_P106258 |
- |
- |
| Agilent Whole Human Genome Oligo Microarray 4x44K |
23817 |
- |
- |
| Agilent Human Genome CGH Microarray 44B |
A_14_P200386 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
29494 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P02984094 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_18_P12123506 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P106258 |
- |
- |
| SurePrint G3 Human GE Microarray G4851A |
A_23_P106258 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16734028 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513956 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513932 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513958 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513950 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513954 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513931 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513939 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513961 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513946 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16734019 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513938 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513919 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513923 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29976016 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16734026 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513936 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29976031 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29976037 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29976038 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513941 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16734024 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513948 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513960 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513952 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513934 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16734018 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513918 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513955 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16734021 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513921 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513929 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513926 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16734034 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513940 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513943 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513926 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16734021 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513958 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513929 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513961 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513923 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513936 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16734019 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513941 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513952 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513954 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513950 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513938 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29976016 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513921 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29976037 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29976038 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513939 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513932 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29976031 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16734034 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16734018 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513940 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513943 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16734028 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513918 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513960 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513931 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513948 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513955 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16734024 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16734026 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513934 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513946 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513956 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09513919 |
- |
- |
| Whole Human Genome Oligo Microarray G4112A |
A_23_P106258 |
- |
- |
| Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P200386 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09513954 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09513934 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09513960 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09513938 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09513955 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09513951 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09513926 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09513919 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09513952 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09513946 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09513956 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09513932 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09513943 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09513936 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09513940 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09513922 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09513958 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09513948 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09513949 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09513920 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09513918 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09513939 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09513941 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P09513980 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16734047 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16734045 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16734048 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16734049 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16734046 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P09513981 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P09513982 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16734050 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16734044 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P200386 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P20161916 |
- |
- |
| Agilent Human Genome CGH Microarray 4x44K |
A_14_P200386 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K G4112F |
A_23_P106258 |
- |
- |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
42371 |
- |
- |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
98482 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P20161916 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P02984090 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P200386 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P02984095 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513922 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513919 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16734036 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16734055 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513990 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513988 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513940 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16734051 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16734031 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513918 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513926 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513972 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513968 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513943 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513951 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16734024 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513954 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513939 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513946 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513981 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16734062 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513960 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513938 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513956 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513985 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16734018 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513934 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513963 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513979 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513931 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513952 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16734039 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513966 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513958 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16734034 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513986 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513955 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513936 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16734028 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513928 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513970 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16734021 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513976 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16734050 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513980 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513948 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09513984 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P02984090 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P02984095 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P200386 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P20161916 |
- |
- |