| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
8093823 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8093825 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8093822 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8093824 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8093821 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
1553912_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04001364.hg.1 |
TC04000050.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04001364.hg.1 |
ENST00000404649.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04001364.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04001365.hg.1 |
TC04000050.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04001365.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04001363.hg.1 |
TC04000050.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04001363.hg.1 |
ENST00000404649.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04001363.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04001359.hg.1 |
TC04000050.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04001359.hg.1 |
ENST00000404649.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04001359.hg.1 |
ENST00000514422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04001359.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04001366.hg.1 |
TC04000050.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04001366.hg.1 |
ENST00000404649.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04001366.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006607.hg.1 |
ENST00000502775.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006607.hg.1 |
ENST00000502775.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006607.hg.1 |
ENST00000502775.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006607.hg.1 |
ENST00000502775.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006607.hg.1 |
ENST00000502775.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006607.hg.1 |
ENST00000514422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006607.hg.1 |
ENST00000514422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006607.hg.1 |
ENST00000514422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006607.hg.1 |
ENST00000514422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006607.hg.1 |
ENST00000514422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006607.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006607.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006607.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006607.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006607.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006607.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006607.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006607.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006607.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04001362.hg.1 |
TC04000050.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04001362.hg.1 |
ENST00000404649.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04001362.hg.1 |
ENST00000514422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04001362.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04001367.hg.1 |
TC04000050.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04001367.hg.1 |
ENST00000404649.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04001367.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04001360.hg.1 |
TC04000050.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04001360.hg.1 |
ENST00000404649.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04001360.hg.1 |
ENST00000514422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04001360.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400145849.hg.1 |
ENST00000502775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400145849.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400145849.hg.1 |
ENST00000502775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400145849.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400145849.hg.1 |
TC0400006607.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400145847.hg.1 |
ENST00000514422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400145847.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400145847.hg.1 |
ENST00000514422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400145847.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400145847.hg.1 |
TC0400006607.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400145851.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400145851.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400145851.hg.1 |
TC0400006607.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400006607.hg.1 |
ENST00000502775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006607.hg.1 |
ENST00000502775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006607.hg.1 |
ENST00000502775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006607.hg.1 |
ENST00000502775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006607.hg.1 |
ENST00000502775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006607.hg.1 |
ENST00000514422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006607.hg.1 |
ENST00000514422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006607.hg.1 |
ENST00000514422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006607.hg.1 |
ENST00000514422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006607.hg.1 |
ENST00000514422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006607.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006607.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006607.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006607.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006607.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006607.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006607.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006607.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006607.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400145846.hg.1 |
ENST00000502775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400145846.hg.1 |
ENST00000514422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400145846.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400145846.hg.1 |
ENST00000502775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400145846.hg.1 |
ENST00000514422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400145846.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400145846.hg.1 |
TC0400006607.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400145848.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400145848.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400145848.hg.1 |
TC0400006607.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400145850.hg.1 |
ENST00000502775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400145850.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400145850.hg.1 |
ENST00000502775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400145850.hg.1 |
NR_040045.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400145850.hg.1 |
TC0400006607.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
1553912_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1553912_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1553912_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1553912_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1553912_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1553912_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1553912_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1553912_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1553912_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1553912_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1553912_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1553912_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1553912_at3 |
- |
- |