| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002796.hg.1 |
ENST00000332192.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002796.hg.1 |
NM_001099852.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002796.hg.1 |
TC01000174.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002799.hg.1 |
ENST00000327795.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002799.hg.1 |
ENST00000332192.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002799.hg.1 |
NM_001099852.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002799.hg.1 |
TC01000174.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006926.hg.1 |
ENST00000316412.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006926.hg.1 |
ENST00000316412.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006926.hg.1 |
ENST00000316412.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006926.hg.1 |
ENST00000316412.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006926.hg.1 |
ENST00000316412.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006926.hg.1 |
ENST00000602960.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006926.hg.1 |
ENST00000602960.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006926.hg.1 |
ENST00000602960.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006926.hg.1 |
ENST00000602960.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006926.hg.1 |
NM_001099852.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006926.hg.1 |
NM_001099852.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006926.hg.1 |
NM_001099852.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006926.hg.1 |
NM_001099852.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006926.hg.1 |
NM_001099852.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002798.hg.1 |
ENST00000327795.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002798.hg.1 |
ENST00000332192.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002798.hg.1 |
NM_001099852.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002798.hg.1 |
TC01000174.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100148268.hg.1 |
ENST00000316412.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100148268.hg.1 |
NM_001099852.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100148268.hg.1 |
ENST00000316412.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100148268.hg.1 |
NM_001099852.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100148268.hg.1 |
TC0100006926.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100148270.hg.1 |
ENST00000316412.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100148270.hg.1 |
ENST00000602960.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100148270.hg.1 |
NM_001099852.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100148270.hg.1 |
ENST00000316412.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100148270.hg.1 |
ENST00000602960.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100148270.hg.1 |
NM_001099852.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100148270.hg.1 |
TC0100006926.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100148271.hg.1 |
ENST00000316412.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100148271.hg.1 |
ENST00000602960.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100148271.hg.1 |
NM_001099852.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100148271.hg.1 |
ENST00000316412.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100148271.hg.1 |
ENST00000602960.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100148271.hg.1 |
NM_001099852.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100148271.hg.1 |
TC0100006926.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100006926.hg.1 |
ENST00000316412.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006926.hg.1 |
ENST00000316412.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006926.hg.1 |
ENST00000316412.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006926.hg.1 |
ENST00000316412.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006926.hg.1 |
ENST00000316412.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006926.hg.1 |
ENST00000602960.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006926.hg.1 |
ENST00000602960.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006926.hg.1 |
ENST00000602960.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006926.hg.1 |
ENST00000602960.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006926.hg.1 |
NM_001099852.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006926.hg.1 |
NM_001099852.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006926.hg.1 |
NM_001099852.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006926.hg.1 |
NM_001099852.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006926.hg.1 |
NM_001099852.2 |
Refseq |