| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018216.hg.1 |
ENST00000374507.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018216.hg.1 |
ENST00000374510.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018216.hg.1 |
ENST00000374511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018216.hg.1 |
ENST00000423740.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018216.hg.1 |
ENST00000461950.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018216.hg.1 |
NM_001003936.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018216.hg.1 |
TC09001474.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
ENST00000374507.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
ENST00000374507.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
ENST00000374507.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
ENST00000374507.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
ENST00000374507.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
ENST00000374510.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
ENST00000374510.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
ENST00000374510.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
ENST00000374510.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
ENST00000374510.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
ENST00000374510.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
ENST00000374510.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
ENST00000374511.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
ENST00000374511.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
ENST00000374511.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
ENST00000374511.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
ENST00000374511.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
ENST00000374511.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
ENST00000374511.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
ENST00000423740.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
ENST00000423740.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
ENST00000423740.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
ENST00000423740.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
ENST00000423740.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
NM_001003936.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
NM_001003936.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
NM_001003936.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
NM_001003936.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
NM_001003936.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
NM_001003936.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
NM_001003936.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
NM_001003936.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
NM_001003936.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
NM_001286946.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
NM_001286946.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
NM_001286946.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
NM_001286946.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
NM_001286946.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
NM_001286946.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
NM_001286946.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
NM_001286946.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
NM_001286947.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
NM_001286947.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
NM_001286947.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
NM_001286947.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
NM_001286947.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011181.hg.1 |
NM_001286947.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018218.hg.1 |
ENST00000374510.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018218.hg.1 |
ENST00000374511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018218.hg.1 |
ENST00000461950.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018218.hg.1 |
NM_001003936.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018218.hg.1 |
TC09001474.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
ENST00000374507.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
ENST00000374507.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
ENST00000374507.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
ENST00000374507.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
ENST00000374507.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
ENST00000374510.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
ENST00000374510.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
ENST00000374510.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
ENST00000374510.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
ENST00000374510.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
ENST00000374510.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
ENST00000374510.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
ENST00000374511.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
ENST00000374511.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
ENST00000374511.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
ENST00000374511.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
ENST00000374511.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
ENST00000374511.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
ENST00000374511.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
ENST00000423740.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
ENST00000423740.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
ENST00000423740.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
ENST00000423740.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
ENST00000423740.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
NM_001003936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
NM_001003936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
NM_001003936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
NM_001003936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
NM_001003936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
NM_001003936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
NM_001003936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
NM_001003936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
NM_001003936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
NM_001286946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
NM_001286946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
NM_001286946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
NM_001286946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
NM_001286946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
NM_001286946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
NM_001286946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
NM_001286946.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
NM_001286947.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
NM_001286947.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
NM_001286947.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
NM_001286947.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
NM_001286947.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011181.hg.1 |
NM_001286947.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900176045.hg.1 |
ENST00000374510.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900176045.hg.1 |
ENST00000374511.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900176045.hg.1 |
NM_001003936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900176045.hg.1 |
ENST00000374510.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900176045.hg.1 |
ENST00000374511.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900176045.hg.1 |
NM_001003936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900176045.hg.1 |
TC0900011181.hg.1 |
Clariom D TC |