| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
XR_040799_at |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
XR_040798_at |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
XR_040800_at |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
XR_017959_at |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8139797 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8139796 |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011211.hg.1 |
ENST00000624231.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011211.hg.1 |
ENST00000624231.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011211.hg.1 |
ENST00000624231.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011211.hg.1 |
ENST00000624231.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000433918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000433918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000433918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000433918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000433918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000433918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000433918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000433918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000433918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000433918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000593670.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000593670.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000593670.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000593670.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000593670.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000593670.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000593670.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000596569.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000596569.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000596569.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000596569.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000596569.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000596569.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000596855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000596855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000596855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000596855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000596855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000596855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000596855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000596855.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000600182.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000600182.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000600182.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000600182.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000600182.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011261.hg.1 |
ENST00000600182.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011211.hg.1 |
ENST00000624231.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011211.hg.1 |
ENST00000624231.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011211.hg.1 |
ENST00000624231.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011211.hg.1 |
ENST00000624231.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000433918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000433918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000433918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000433918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000433918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000433918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000433918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000433918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000433918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000433918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587704.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000587736.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000593670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000593670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000593670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000593670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000593670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000593670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000593670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000594855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000596569.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000596569.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000596569.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000596569.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000596569.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000596569.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000596855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000596855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000596855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000596855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000596855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000596855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000596855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000596855.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000600182.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000600182.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000600182.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000600182.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000600182.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011261.hg.1 |
ENST00000600182.2 |
Ensembl |