| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
NM_024114_at |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7939915 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7950972 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7943027 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7950974 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7950976 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7943026 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7950975 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7939914 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7943028 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7943030 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7939913 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
7939912 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
7950971 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
7943025 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
220534_PM_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11735773_x_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11735773_PM_x_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011921.hg.1 |
ENST00000532014.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011921.hg.1 |
ENST00000532014.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011921.hg.1 |
ENST00000532014.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011921.hg.1 |
ENST00000532014.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011921.hg.1 |
ENST00000532014.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008721.hg.1 |
ENST00000525732.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008721.hg.1 |
ENST00000525732.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008721.hg.1 |
ENST00000525732.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008721.hg.1 |
ENST00000525732.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008721.hg.1 |
ENST00000525732.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011920.hg.1 |
ENST00000524374.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011920.hg.1 |
ENST00000524374.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011920.hg.1 |
ENST00000524374.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011920.hg.1 |
ENST00000524374.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011920.hg.1 |
ENST00000524374.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011920.hg.1 |
ENST00000524374.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008724.hg.1 |
ENST00000526396.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008724.hg.1 |
ENST00000526396.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008724.hg.1 |
ENST00000526396.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008724.hg.1 |
ENST00000526396.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008724.hg.1 |
ENST00000526396.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008724.hg.1 |
ENST00000623787.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008724.hg.1 |
ENST00000623787.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008724.hg.1 |
ENST00000623787.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008724.hg.1 |
ENST00000623787.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008724.hg.1 |
ENST00000623787.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008724.hg.1 |
ENST00000623787.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008724.hg.1 |
ENST00000623787.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008724.hg.1 |
ENST00000623787.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008724.hg.1 |
NM_001206627.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008724.hg.1 |
NM_001206627.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008724.hg.1 |
NM_001206627.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008724.hg.1 |
NM_001206627.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008724.hg.1 |
NM_001206627.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008724.hg.1 |
NM_001206627.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011921.hg.1 |
ENST00000532014.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011921.hg.1 |
ENST00000532014.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011921.hg.1 |
ENST00000532014.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011921.hg.1 |
ENST00000532014.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011921.hg.1 |
ENST00000532014.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000420869.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000530311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
ENST00000605881.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011917.hg.1 |
NM_001105522.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011920.hg.1 |
ENST00000524374.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011920.hg.1 |
ENST00000524374.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011920.hg.1 |
ENST00000524374.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011920.hg.1 |
ENST00000524374.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011920.hg.1 |
ENST00000524374.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011920.hg.1 |
ENST00000524374.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008721.hg.1 |
ENST00000525732.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008721.hg.1 |
ENST00000525732.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008721.hg.1 |
ENST00000525732.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008721.hg.1 |
ENST00000525732.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008721.hg.1 |
ENST00000525732.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008724.hg.1 |
ENST00000526396.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008724.hg.1 |
ENST00000526396.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008724.hg.1 |
ENST00000526396.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008724.hg.1 |
ENST00000526396.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008724.hg.1 |
ENST00000526396.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008724.hg.1 |
ENST00000623787.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008724.hg.1 |
ENST00000623787.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008724.hg.1 |
ENST00000623787.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008724.hg.1 |
ENST00000623787.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008724.hg.1 |
ENST00000623787.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008724.hg.1 |
ENST00000623787.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008724.hg.1 |
ENST00000623787.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008724.hg.1 |
ENST00000623787.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008724.hg.1 |
NM_001206627.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008724.hg.1 |
NM_001206627.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008724.hg.1 |
NM_001206627.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008724.hg.1 |
NM_001206627.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008724.hg.1 |
NM_001206627.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008724.hg.1 |
NM_001206627.1 |
Refseq |
| Human Genome U133 Plus 2.0 Array |
220534_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
220534_at |
- |
- |
| Human Genome U133A 2.0 Array |
220534_PM_at |
- |
- |
| Human Genome U133A 2.0 Array |
220534_at |
- |
- |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) |
NM_024114_at |
- |
- |
| AffymetrixPathwork Diagnostics Pathchip |
pw502349 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
220534_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
220534_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
220534_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
220534_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
220534_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
220534_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
220534_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
220534_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
220534_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
220534_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
220534_at8 |
- |
- |
| Human Genome U133 Set |
220534_at |
- |
- |
| Human Genome U133 Set |
220534_PM_at |
- |
- |