| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
8005445 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8005444 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
1564383_PM_s_at |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
1557895_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010058.hg.1 |
ENST00000563569.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010058.hg.1 |
ENST00000563569.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010058.hg.1 |
ENST00000578659.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010058.hg.1 |
ENST00000578659.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010058.hg.1 |
ENST00000578659.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010058.hg.1 |
ENST00000579603.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010058.hg.1 |
ENST00000579603.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010058.hg.1 |
ENST00000579603.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010058.hg.1 |
ENST00000583797.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010058.hg.1 |
ENST00000583797.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010058.hg.1 |
ENST00000584274.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007121.hg.1 |
ENST00000577684.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007121.hg.1 |
ENST00000577684.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007121.hg.1 |
ENST00000577684.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007121.hg.1 |
ENST00000577684.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007121.hg.1 |
ENST00000577684.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007121.hg.1 |
NR_104343.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007121.hg.1 |
NR_104343.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007121.hg.1 |
NR_104343.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007121.hg.1 |
NR_104343.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007121.hg.1 |
NR_104343.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700150335.hg.1 |
ENST00000577684.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700150335.hg.1 |
NR_104343.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700150335.hg.1 |
ENST00000577684.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700150335.hg.1 |
NR_104343.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700150335.hg.1 |
TC1700007121.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1700007121.hg.1 |
ENST00000577684.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007121.hg.1 |
ENST00000577684.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007121.hg.1 |
ENST00000577684.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007121.hg.1 |
ENST00000577684.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007121.hg.1 |
ENST00000577684.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007121.hg.1 |
NR_104343.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007121.hg.1 |
NR_104343.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007121.hg.1 |
NR_104343.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007121.hg.1 |
NR_104343.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007121.hg.1 |
NR_104343.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010058.hg.1 |
ENST00000342824.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010058.hg.1 |
ENST00000563569.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010058.hg.1 |
ENST00000563569.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010058.hg.1 |
ENST00000578659.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010058.hg.1 |
ENST00000578659.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010058.hg.1 |
ENST00000578659.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010058.hg.1 |
ENST00000579603.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010058.hg.1 |
ENST00000579603.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010058.hg.1 |
ENST00000579603.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010058.hg.1 |
ENST00000583797.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010058.hg.1 |
ENST00000583797.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010058.hg.1 |
ENST00000584274.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700150328.hg.1 |
ENST00000577684.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700150328.hg.1 |
NR_104343.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700150328.hg.1 |
ENST00000577684.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700150328.hg.1 |
NR_104343.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700150328.hg.1 |
TC1700007121.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700150332.hg.1 |
ENST00000577684.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700150332.hg.1 |
NR_104343.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700150332.hg.1 |
ENST00000577684.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700150332.hg.1 |
NR_104343.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700150332.hg.1 |
TC1700007121.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700150334.hg.1 |
ENST00000577684.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700150334.hg.1 |
NR_104343.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700150334.hg.1 |
ENST00000577684.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700150334.hg.1 |
NR_104343.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700150334.hg.1 |
TC1700007121.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700150329.hg.1 |
ENST00000577684.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700150329.hg.1 |
NR_104343.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700150329.hg.1 |
ENST00000577684.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700150329.hg.1 |
NR_104343.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700150329.hg.1 |
TC1700007121.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
1557895_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1557895_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1564383_s_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1564383_PM_s_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1564383_s_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557895_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557895_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1564383_s_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557895_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557895_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1564383_s_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1564383_s_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557895_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1564383_s_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1564383_s_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557895_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1564383_s_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557895_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557895_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557895_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1564383_s_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557895_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1564383_s_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1564383_s_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1564383_s_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557895_at4 |
- |
- |
| Human Genome U95 Set |
61766_PM_at |
- |
- |
| Human Genome U95 Set |
61766_at |
- |
- |