| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
8067828 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8067830 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8067832 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8067831 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8067833 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8067826 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8067829 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8067827 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8067825 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
1552895_PM_a_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11755711_PM_s_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11755711_s_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11746501_x_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11746501_PM_x_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006444.hg.1 |
ENST00000448473.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006444.hg.1 |
ENST00000448473.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006444.hg.1 |
ENST00000448473.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006444.hg.1 |
ENST00000448473.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006444.hg.1 |
ENST00000448473.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21000037.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21000037.hg.1 |
TC21000013.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21000036.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21000036.hg.1 |
TC21000013.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000447861.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000447861.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000447861.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000447861.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000447861.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000447861.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000447861.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000447861.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000447861.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000447861.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000471407.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000471407.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000471407.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000471407.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000471407.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000507941.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000507941.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000507941.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000507941.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
ENST00000507941.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21000038.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21000038.hg.1 |
TC21000013.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014344.hg.1 |
ENST00000452011.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014344.hg.1 |
ENST00000452011.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014344.hg.1 |
ENST00000452011.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014344.hg.1 |
ENST00000452011.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21000034.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21000034.hg.1 |
TC21000013.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21000033.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21000033.hg.1 |
TC21000013.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21000035.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21000035.hg.1 |
TC21000013.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0200014344.hg.1 |
ENST00000452011.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014344.hg.1 |
ENST00000452011.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014344.hg.1 |
ENST00000452011.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014344.hg.1 |
ENST00000452011.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100144599.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100144599.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100144599.hg.1 |
TC2100006541.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100144594.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100144594.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100144594.hg.1 |
TC2100006541.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100144598.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100144598.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100144598.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100144598.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100144598.hg.1 |
TC2100006541.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100144603.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100144603.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100144603.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100144603.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100144603.hg.1 |
TC2100006541.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100144607.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100144607.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100144607.hg.1 |
TC2100006541.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC2200006444.hg.1 |
ENST00000448473.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006444.hg.1 |
ENST00000448473.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006444.hg.1 |
ENST00000448473.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006444.hg.1 |
ENST00000448473.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006444.hg.1 |
ENST00000448473.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100144602.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100144602.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100144602.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100144602.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100144602.hg.1 |
TC2100006541.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000447861.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000447861.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000447861.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000447861.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000447861.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000447861.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000447861.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000447861.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000447861.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000447861.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000471407.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000471407.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000471407.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000471407.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000471407.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000507941.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000507941.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000507941.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000507941.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
ENST00000507941.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006541.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100144592.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100144592.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100144592.hg.1 |
TC2100006541.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100144593.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100144593.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100144593.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100144593.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100144593.hg.1 |
TC2100006541.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100144601.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100144601.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100144601.hg.1 |
ENST00000435744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100144601.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100144601.hg.1 |
TC2100006541.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100144606.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100144606.hg.1 |
NR_026916.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100144606.hg.1 |
TC2100006541.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
1552895_a_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1552895_PM_a_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1552895_a_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1552895_a_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1552895_a_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1552895_a_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1552895_a_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1552895_a_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1552895_a_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1552895_a_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1552895_a_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1552895_a_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1552895_a_at4 |
- |
- |
| Human Genome U95 Set |
69934_PM_at |
- |
- |
| Human Genome U95 Set |
69934_at |
- |
- |
| Human Genome U95 Set |
90042_PM_f_at |
- |
- |
| Human Genome U95 Set |
90042_f_at |
- |
- |