| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019432.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019432.hg.1 |
ENST00000525746.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019432.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019432.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019432.hg.1 |
TC06001595.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019441.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019441.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019441.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019441.hg.1 |
TC06001595.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
ENST00000525912.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
ENST00000525746.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
ENST00000525746.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
ENST00000525746.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
ENST00000525912.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC06001595.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019440.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019440.hg.1 |
ENST00000525912.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019440.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019440.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019440.hg.1 |
TC06001595.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019433.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019433.hg.1 |
ENST00000525746.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019433.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019433.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019433.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019433.hg.1 |
TC06001595.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019443.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019443.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019443.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019443.hg.1 |
TC06001595.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019430.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019430.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019430.hg.1 |
TC06001595.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019431.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019431.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019431.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019431.hg.1 |
TC06001595.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600179316.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179316.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179316.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179316.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179316.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179316.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179316.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179316.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179316.hg.1 |
TC0600011569.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600179318.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179318.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179318.hg.1 |
ENST00000525746.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179318.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179318.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179318.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179318.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179318.hg.1 |
ENST00000525746.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179318.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179318.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179318.hg.1 |
TC0600011569.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600179314.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179314.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179314.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179314.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179314.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179314.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179314.hg.1 |
TC0600011569.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600179330.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179330.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179330.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179330.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179330.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179330.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179330.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179330.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179330.hg.1 |
TC0600011569.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000525746.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000525746.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000525746.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000525746.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000525912.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000525912.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011569.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179317.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179317.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179317.hg.1 |
ENST00000525746.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179317.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179317.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179317.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179317.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179317.hg.1 |
ENST00000525746.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179317.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179317.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179317.hg.1 |
TC0600011569.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600179320.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179320.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179320.hg.1 |
ENST00000525746.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179320.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179320.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179320.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179320.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179320.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179320.hg.1 |
ENST00000525746.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179320.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179320.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179320.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179320.hg.1 |
TC0600011569.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600179326.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179326.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179326.hg.1 |
ENST00000525912.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179326.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179326.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179326.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179326.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179326.hg.1 |
ENST00000525912.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179326.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179326.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179326.hg.1 |
TC0600011569.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600179327.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179327.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179327.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179327.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179327.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179327.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179327.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179327.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179327.hg.1 |
TC0600011569.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600179313.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179313.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179313.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179313.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179313.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179313.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179313.hg.1 |
TC0600011569.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
1557276_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1557277_PM_a_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1557277_a_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1556675_s_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1557276_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1556675_PM_s_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557277_a_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557276_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556675_s_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557277_a_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557277_a_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557276_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556675_s_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557277_a_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556675_s_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557276_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556675_s_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556675_s_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557277_a_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557276_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556675_s_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557277_a_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557276_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557276_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557277_a_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557276_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556675_s_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557277_a_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557277_a_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556675_s_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557276_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557276_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556675_s_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556675_s_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557277_a_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557277_a_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556675_s_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557276_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557276_at2 |
- |
- |