Search
Search

HSPE1P13| Gene Symbol: HSPE1P13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC047033.1 | - | 1 | 252 | 99 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Rho guanine nucleotide exchange factor 33 | ARHGEF33 | -122.4kb | Anti-sense | Chr. 2 - 38889875 - 38975454 on NCBI Build GRCh38 | |
| Downstream | cyclin dependent kinase like 4 | CDKL4 | 69.3kb | Sense | Chr. 2 - 39167971 - 39247120 on NCBI Build GRCh38 |
Set Membership: |
|