| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
7977665 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7977670 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7977660 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8047061 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7977668 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8047060 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7977663 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
7912552 |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018170.hg.1 |
ENST00000617807.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018170.hg.1 |
ENST00000617807.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018170.hg.1 |
ENST00000617807.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018170.hg.1 |
ENST00000617807.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018170.hg.1 |
NM_001146181.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018170.hg.1 |
NM_001146181.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018170.hg.1 |
NM_001302551.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01034194.hg.1 |
NM_001136561.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01034194.hg.1 |
TC01002221.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01034195.hg.1 |
NM_001136561.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01034195.hg.1 |
TC01002221.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01034193.hg.1 |
NM_001136561.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01034193.hg.1 |
TC01002221.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100195547.hg.1 |
ENST00000621994.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195547.hg.1 |
NM_001136561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195547.hg.1 |
ENST00000621994.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195547.hg.1 |
NM_001136561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195547.hg.1 |
TC0100012938.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100236970.hg.1 |
ENST00000617807.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100236970.hg.1 |
NM_001146181.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100236970.hg.1 |
NM_001302551.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100236970.hg.1 |
ENST00000617807.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100236970.hg.1 |
NM_001146181.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100236970.hg.1 |
NM_001302551.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100236970.hg.1 |
TC0100018170.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100236968.hg.1 |
ENST00000617807.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100236968.hg.1 |
NM_001146181.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100236968.hg.1 |
ENST00000617807.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100236968.hg.1 |
NM_001146181.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100236968.hg.1 |
TC0100018170.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100018170.hg.1 |
ENST00000617807.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018170.hg.1 |
ENST00000617807.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018170.hg.1 |
ENST00000617807.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018170.hg.1 |
ENST00000617807.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018170.hg.1 |
NM_001146181.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018170.hg.1 |
NM_001146181.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018170.hg.1 |
NM_001302551.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195549.hg.1 |
ENST00000621994.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195549.hg.1 |
NM_001136561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195549.hg.1 |
ENST00000621994.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195549.hg.1 |
NM_001136561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195549.hg.1 |
TC0100012938.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100195548.hg.1 |
ENST00000621994.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195548.hg.1 |
NM_001136561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195548.hg.1 |
ENST00000621994.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195548.hg.1 |
NM_001136561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195548.hg.1 |
TC0100012938.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
216302_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
216302_PM_at |
- |
- |
| Human Genome U133A 2.0 Array |
216302_PM_at |
- |
- |
| Human Genome U133A 2.0 Array |
216302_at |
- |
- |
| Human Genome U95 Set |
32408_PM_s_at |
- |
- |
| Human Genome U95 Set |
32408_s_at |
- |
- |
| Human Genome U133 Set |
216302_PM_at |
- |
- |
| Human Genome U133 Set |
216302_at |
- |
- |