| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17005513.hg.1 |
NR_038847.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17005513.hg.1 |
TC17000470.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17005514.hg.1 |
NR_038847.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17005514.hg.1 |
TC17000470.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007727.hg.1 |
ENST00000583195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007727.hg.1 |
ENST00000583195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007727.hg.1 |
ENST00000583195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007727.hg.1 |
ENST00000583195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007727.hg.1 |
NR_038847.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007727.hg.1 |
NR_038847.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007727.hg.1 |
NR_038847.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007727.hg.1 |
NR_038847.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007727.hg.1 |
NR_038847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700155756.hg.1 |
ENST00000583195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700155756.hg.1 |
NR_038847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700155756.hg.1 |
ENST00000583195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700155756.hg.1 |
NR_038847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700155756.hg.1 |
TC1700007727.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700155758.hg.1 |
ENST00000583195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700155758.hg.1 |
NR_038847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700155758.hg.1 |
ENST00000583195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700155758.hg.1 |
NR_038847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700155758.hg.1 |
TC1700007727.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1700007727.hg.1 |
ENST00000583195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007727.hg.1 |
ENST00000583195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007727.hg.1 |
ENST00000583195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007727.hg.1 |
ENST00000583195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007727.hg.1 |
NR_038847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007727.hg.1 |
NR_038847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007727.hg.1 |
NR_038847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007727.hg.1 |
NR_038847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007727.hg.1 |
NR_038847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700155761.hg.1 |
NR_038847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700155761.hg.1 |
NR_038847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700155761.hg.1 |
TC1700007727.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700155760.hg.1 |
ENST00000583195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700155760.hg.1 |
NR_038847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700155760.hg.1 |
ENST00000583195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700155760.hg.1 |
NR_038847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700155760.hg.1 |
TC1700007727.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700155759.hg.1 |
ENST00000583195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700155759.hg.1 |
NR_038847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700155759.hg.1 |
ENST00000583195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700155759.hg.1 |
NR_038847.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700155759.hg.1 |
TC1700007727.hg.1 |
Clariom D TC |