| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012304.hg.1 |
ENST00000519550.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012304.hg.1 |
ENST00000519550.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012304.hg.1 |
ENST00000519550.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012304.hg.1 |
ENST00000519550.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012304.hg.1 |
ENST00000519550.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012304.hg.1 |
ENST00000519550.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012304.hg.1 |
NR_038875.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012304.hg.1 |
NR_038875.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012304.hg.1 |
NR_038875.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012304.hg.1 |
NR_038875.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012304.hg.1 |
NR_038875.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012304.hg.1 |
NR_038875.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08005960.hg.1 |
ENST00000519550.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08005960.hg.1 |
NR_038875.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08005960.hg.1 |
TC08000429.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08005959.hg.1 |
ENST00000519550.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08005959.hg.1 |
NR_038875.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08005959.hg.1 |
TC08000429.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0800012304.hg.1 |
ENST00000519550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012304.hg.1 |
ENST00000519550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012304.hg.1 |
ENST00000519550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012304.hg.1 |
ENST00000519550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012304.hg.1 |
ENST00000519550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012304.hg.1 |
ENST00000519550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012304.hg.1 |
NR_038875.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012304.hg.1 |
NR_038875.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012304.hg.1 |
NR_038875.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012304.hg.1 |
NR_038875.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012304.hg.1 |
NR_038875.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012304.hg.1 |
NR_038875.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800185376.hg.1 |
ENST00000519550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800185376.hg.1 |
NR_038875.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800185376.hg.1 |
ENST00000519550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800185376.hg.1 |
NR_038875.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800185376.hg.1 |
TC0800012304.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800185379.hg.1 |
ENST00000519550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800185379.hg.1 |
NR_038875.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800185379.hg.1 |
ENST00000519550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800185379.hg.1 |
NR_038875.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800185379.hg.1 |
TC0800012304.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800185380.hg.1 |
ENST00000519550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800185380.hg.1 |
NR_038875.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800185380.hg.1 |
ENST00000519550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800185380.hg.1 |
NR_038875.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800185380.hg.1 |
TC0800012304.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800185378.hg.1 |
ENST00000519550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800185378.hg.1 |
NR_038875.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800185378.hg.1 |
ENST00000519550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800185378.hg.1 |
NR_038875.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800185378.hg.1 |
TC0800012304.hg.1 |
Clariom D TC |