| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008586.hg.1 |
ENST00000453380.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008586.hg.1 |
NM_001271560.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008586.hg.1 |
NM_001271560.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008586.hg.1 |
NM_001271560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158307.hg.1 |
NM_001271560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158307.hg.1 |
NM_001271560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158307.hg.1 |
TC0X00008586.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00158309.hg.1 |
NM_001271560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158309.hg.1 |
NM_001271560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158309.hg.1 |
TC0X00008586.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00158308.hg.1 |
ENST00000453380.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158308.hg.1 |
NM_001271560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158308.hg.1 |
ENST00000453380.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158308.hg.1 |
NM_001271560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158308.hg.1 |
TC0X00008586.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00008586.hg.1 |
ENST00000453380.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008586.hg.1 |
NM_001271560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008586.hg.1 |
NM_001271560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008586.hg.1 |
NM_001271560.1 |
Refseq |