| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008455.hg.1 |
ENST00000342537.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008455.hg.1 |
ENST00000342537.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008455.hg.1 |
ENST00000342537.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008455.hg.1 |
ENST00000342537.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008455.hg.1 |
ENST00000555360.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008455.hg.1 |
ENST00000555360.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008455.hg.1 |
NM_001137601.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008455.hg.1 |
NM_001137601.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008455.hg.1 |
NM_001137601.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008455.hg.1 |
ENST00000342537.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008455.hg.1 |
ENST00000342537.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008455.hg.1 |
ENST00000342537.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008455.hg.1 |
ENST00000342537.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008455.hg.1 |
ENST00000555360.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008455.hg.1 |
ENST00000555360.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008455.hg.1 |
NM_001137601.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008455.hg.1 |
NM_001137601.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008455.hg.1 |
NM_001137601.2 |
Refseq |