| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007050.hg.1 |
ENST00000600179.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007050.hg.1 |
NM_001310155.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007050.hg.1 |
NM_001310155.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007050.hg.1 |
NM_001310155.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007063.hg.1 |
ENST00000434336.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007063.hg.1 |
NM_001310138.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007063.hg.1 |
NM_001310138.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013057.hg.1 |
ENST00000624031.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1_KI270713v1_random00006432.hg.1 |
ENST00000601199.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1_KI270713v1_random00006432.hg.1 |
NM_001282321.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007050.hg.1 |
ENST00000600179.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007050.hg.1 |
NM_001310155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007050.hg.1 |
NM_001310155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007050.hg.1 |
NM_001310155.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013057.hg.1 |
ENST00000624031.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1_KI270713v1_random00006432.hg.1 |
ENST00000601199.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1_KI270713v1_random00006432.hg.1 |
NM_001282321.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007063.hg.1 |
ENST00000434336.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007063.hg.1 |
NM_001310138.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007063.hg.1 |
NM_001310138.1 |
Refseq |