| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009522.hg.1 |
ENST00000519147.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009522.hg.1 |
ENST00000519147.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009522.hg.1 |
ENST00000519147.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009522.hg.1 |
ENST00000522213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009522.hg.1 |
ENST00000522213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009522.hg.1 |
ENST00000522213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009522.hg.1 |
ENST00000522213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009522.hg.1 |
ENST00000522213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009522.hg.1 |
ENST00000522213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009522.hg.1 |
ENST00000522213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009522.hg.1 |
ENST00000522213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006952.hg.1 |
ENST00000507936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006952.hg.1 |
ENST00000507936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006952.hg.1 |
ENST00000507936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006952.hg.1 |
ENST00000507936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006952.hg.1 |
ENST00000507986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006952.hg.1 |
ENST00000507986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006952.hg.1 |
ENST00000507986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006952.hg.1 |
ENST00000512559.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006952.hg.1 |
ENST00000512559.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006952.hg.1 |
ENST00000512559.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006952.hg.1 |
ENST00000512559.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006952.hg.1 |
ENST00000512559.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006952.hg.1 |
ENST00000512559.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006952.hg.1 |
ENST00000512559.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006952.hg.1 |
ENST00000512559.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006952.hg.1 |
ENST00000512559.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006952.hg.1 |
NR_131245.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006952.hg.1 |
NR_131245.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006952.hg.1 |
NR_131245.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006952.hg.1 |
NR_131245.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006952.hg.1 |
NR_131245.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006952.hg.1 |
NR_131245.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006952.hg.1 |
NR_131245.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006952.hg.1 |
NR_131245.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006952.hg.1 |
NR_131245.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006952.hg.1 |
ENST00000507936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006952.hg.1 |
ENST00000507936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006952.hg.1 |
ENST00000507936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006952.hg.1 |
ENST00000507936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006952.hg.1 |
ENST00000507986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006952.hg.1 |
ENST00000507986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006952.hg.1 |
ENST00000507986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006952.hg.1 |
ENST00000512559.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006952.hg.1 |
ENST00000512559.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006952.hg.1 |
ENST00000512559.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006952.hg.1 |
ENST00000512559.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006952.hg.1 |
ENST00000512559.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006952.hg.1 |
ENST00000512559.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006952.hg.1 |
ENST00000512559.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006952.hg.1 |
ENST00000512559.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006952.hg.1 |
ENST00000512559.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006952.hg.1 |
NR_131245.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006952.hg.1 |
NR_131245.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006952.hg.1 |
NR_131245.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006952.hg.1 |
NR_131245.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006952.hg.1 |
NR_131245.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006952.hg.1 |
NR_131245.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006952.hg.1 |
NR_131245.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006952.hg.1 |
NR_131245.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006952.hg.1 |
NR_131245.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009522.hg.1 |
ENST00000519147.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009522.hg.1 |
ENST00000519147.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009522.hg.1 |
ENST00000519147.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009522.hg.1 |
ENST00000522213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009522.hg.1 |
ENST00000522213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009522.hg.1 |
ENST00000522213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009522.hg.1 |
ENST00000522213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009522.hg.1 |
ENST00000522213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009522.hg.1 |
ENST00000522213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009522.hg.1 |
ENST00000522213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009522.hg.1 |
ENST00000522213.1 |
Ensembl |