| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010196.hg.1 |
ENST00000513157.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010196.hg.1 |
ENST00000513157.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010196.hg.1 |
ENST00000513157.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010196.hg.1 |
ENST00000513157.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010196.hg.1 |
ENST00000513157.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010196.hg.1 |
ENST00000513157.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010196.hg.1 |
ENST00000513157.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010196.hg.1 |
ENST00000513157.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010196.hg.1 |
ENST00000513157.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010196.hg.1 |
NR_104625.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010196.hg.1 |
NR_104625.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010196.hg.1 |
NR_104625.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010196.hg.1 |
NR_104625.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010196.hg.1 |
NR_104625.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010196.hg.1 |
NR_104625.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010196.hg.1 |
NR_104625.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010196.hg.1 |
NR_104625.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010196.hg.1 |
NR_104625.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010196.hg.1 |
ENST00000513157.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010196.hg.1 |
ENST00000513157.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010196.hg.1 |
ENST00000513157.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010196.hg.1 |
ENST00000513157.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010196.hg.1 |
ENST00000513157.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010196.hg.1 |
ENST00000513157.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010196.hg.1 |
ENST00000513157.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010196.hg.1 |
ENST00000513157.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010196.hg.1 |
ENST00000513157.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010196.hg.1 |
NR_104625.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010196.hg.1 |
NR_104625.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010196.hg.1 |
NR_104625.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010196.hg.1 |
NR_104625.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010196.hg.1 |
NR_104625.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010196.hg.1 |
NR_104625.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010196.hg.1 |
NR_104625.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010196.hg.1 |
NR_104625.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010196.hg.1 |
NR_104625.1 |
Refseq |