Search
Search

MARCH3| Gene Symbol: MARCH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178450.4 | NP_848545.1 | 5 | 4070 | 98 | |
| XM_011543128.2 | XP_011541430.1 | 4 | 3807 | 98 | ||
| XM_011543129.1 | XP_011541431.1 | 6 | 3935 | 98 | ||
| XM_011543130.1 | XP_011541432.1 | 6 | 3840 | 98 | ||
| XM_011543131.2 | XP_011541433.1 | 6 | 4069 | 98 | ||
| XM_017009014.1 | XP_016864503.1 | 4 | 4258 | 98 | ||
| GenBank mRNA | AK000103.1 | - | 1 | 1624 | 98 | |
Set Membership: |
|