| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007433.hg.1 |
ENST00000608605.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007433.hg.1 |
ENST00000608605.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007433.hg.1 |
ENST00000608605.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007433.hg.1 |
ENST00000608605.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007433.hg.1 |
ENST00000608605.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007433.hg.1 |
ENST00000608605.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007433.hg.1 |
ENST00000608605.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007433.hg.1 |
ENST00000609204.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007433.hg.1 |
ENST00000609204.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007433.hg.1 |
ENST00000609204.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007725.hg.1 |
ENST00000578803.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007725.hg.1 |
ENST00000578803.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007725.hg.1 |
ENST00000578803.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007725.hg.1 |
ENST00000578803.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007725.hg.1 |
ENST00000578803.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007725.hg.1 |
ENST00000578803.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007725.hg.1 |
ENST00000579781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007725.hg.1 |
ENST00000579781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007725.hg.1 |
ENST00000579781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007725.hg.1 |
ENST00000579781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007725.hg.1 |
ENST00000579781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007725.hg.1 |
NR_120419.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007725.hg.1 |
NR_120419.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007725.hg.1 |
NR_120419.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007725.hg.1 |
NR_120419.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007725.hg.1 |
NR_120419.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007725.hg.1 |
NR_120419.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007433.hg.1 |
ENST00000608605.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007433.hg.1 |
ENST00000608605.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007433.hg.1 |
ENST00000608605.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007433.hg.1 |
ENST00000608605.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007433.hg.1 |
ENST00000608605.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007433.hg.1 |
ENST00000608605.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007433.hg.1 |
ENST00000608605.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007433.hg.1 |
ENST00000609204.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007433.hg.1 |
ENST00000609204.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007433.hg.1 |
ENST00000609204.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007725.hg.1 |
ENST00000578803.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007725.hg.1 |
ENST00000578803.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007725.hg.1 |
ENST00000578803.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007725.hg.1 |
ENST00000578803.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007725.hg.1 |
ENST00000578803.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007725.hg.1 |
ENST00000578803.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007725.hg.1 |
ENST00000579781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007725.hg.1 |
ENST00000579781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007725.hg.1 |
ENST00000579781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007725.hg.1 |
ENST00000579781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007725.hg.1 |
ENST00000579781.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007725.hg.1 |
NR_120419.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007725.hg.1 |
NR_120419.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007725.hg.1 |
NR_120419.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007725.hg.1 |
NR_120419.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007725.hg.1 |
NR_120419.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007725.hg.1 |
NR_120419.1 |
Refseq |