| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800006492.hg.1 |
ENST00000320248.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800006492.hg.1 |
ENST00000320248.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800006492.hg.1 |
ENST00000320248.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800006492.hg.1 |
ENST00000320248.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800006492.hg.1 |
NM_014867.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800006492.hg.1 |
NM_014867.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800006492.hg.1 |
NM_014867.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800006492.hg.1 |
NM_014867.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800006492.hg.1 |
ENST00000320248.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800006492.hg.1 |
ENST00000320248.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800006492.hg.1 |
ENST00000320248.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800006492.hg.1 |
ENST00000320248.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800006492.hg.1 |
NM_014867.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800006492.hg.1 |
NM_014867.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800006492.hg.1 |
NM_014867.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800006492.hg.1 |
NM_014867.2 |
Refseq |