| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009986.hg.1 |
ENST00000424165.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009986.hg.1 |
ENST00000424165.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009986.hg.1 |
ENST00000424165.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009986.hg.1 |
ENST00000424165.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009986.hg.1 |
ENST00000424165.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009986.hg.1 |
ENST00000448425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009986.hg.1 |
ENST00000448425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009986.hg.1 |
NR_047021.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009986.hg.1 |
NR_047021.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009986.hg.1 |
NR_047021.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009986.hg.1 |
NR_047021.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009986.hg.1 |
NR_047021.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009986.hg.1 |
ENST00000424165.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009986.hg.1 |
ENST00000424165.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009986.hg.1 |
ENST00000424165.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009986.hg.1 |
ENST00000424165.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009986.hg.1 |
ENST00000424165.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009986.hg.1 |
ENST00000448425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009986.hg.1 |
ENST00000448425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009986.hg.1 |
NR_047021.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009986.hg.1 |
NR_047021.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009986.hg.1 |
NR_047021.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009986.hg.1 |
NR_047021.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009986.hg.1 |
NR_047021.1 |
Refseq |