| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011941.hg.1 |
ENST00000419373.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011941.hg.1 |
ENST00000419373.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011941.hg.1 |
ENST00000419373.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011941.hg.1 |
ENST00000419373.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011941.hg.1 |
ENST00000419373.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011941.hg.1 |
ENST00000419373.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011941.hg.1 |
ENST00000419373.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011941.hg.1 |
ENST00000419373.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011941.hg.1 |
NR_121650.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011941.hg.1 |
NR_121650.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011941.hg.1 |
NR_121650.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011941.hg.1 |
NR_121650.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011941.hg.1 |
NR_121650.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011941.hg.1 |
NR_121650.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011941.hg.1 |
NR_121650.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011941.hg.1 |
NR_121650.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011941.hg.1 |
NR_121651.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011941.hg.1 |
NR_121651.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011941.hg.1 |
NR_121651.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011941.hg.1 |
NR_121651.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011941.hg.1 |
NR_121651.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008989.hg.1 |
ENST00000584078.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008989.hg.1 |
ENST00000584078.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008989.hg.1 |
ENST00000584078.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011941.hg.1 |
ENST00000419373.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011941.hg.1 |
ENST00000419373.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011941.hg.1 |
ENST00000419373.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011941.hg.1 |
ENST00000419373.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011941.hg.1 |
ENST00000419373.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011941.hg.1 |
ENST00000419373.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011941.hg.1 |
ENST00000419373.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011941.hg.1 |
ENST00000419373.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011941.hg.1 |
NR_121650.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011941.hg.1 |
NR_121650.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011941.hg.1 |
NR_121650.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011941.hg.1 |
NR_121650.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011941.hg.1 |
NR_121650.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011941.hg.1 |
NR_121650.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011941.hg.1 |
NR_121650.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011941.hg.1 |
NR_121650.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011941.hg.1 |
NR_121651.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011941.hg.1 |
NR_121651.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011941.hg.1 |
NR_121651.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011941.hg.1 |
NR_121651.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011941.hg.1 |
NR_121651.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008989.hg.1 |
ENST00000584078.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008989.hg.1 |
ENST00000584078.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008989.hg.1 |
ENST00000584078.1 |
Ensembl |