| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015618.hg.1 |
ENST00000497012.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013217.hg.1 |
ENST00000455962.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015492.hg.1 |
ENST00000604437.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015492.hg.1 |
ENST00000604437.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015492.hg.1 |
ENST00000621347.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015492.hg.1 |
NR_003242.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015492.hg.1 |
NR_003242.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015492.hg.1 |
NR_003242.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009808.hg.1 |
ENST00000450970.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009709.hg.1 |
ENST00000476786.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009732.hg.1 |
ENST00000443612.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009732.hg.1 |
ENST00000443612.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009808.hg.1 |
ENST00000450970.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013217.hg.1 |
ENST00000455962.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009709.hg.1 |
ENST00000476786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015492.hg.1 |
ENST00000604437.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015492.hg.1 |
ENST00000604437.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015492.hg.1 |
ENST00000621347.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015492.hg.1 |
NR_003242.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015492.hg.1 |
NR_003242.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015492.hg.1 |
NR_003242.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015618.hg.1 |
ENST00000497012.2 |
Ensembl |