| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010898.hg.1 |
ENST00000580257.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010898.hg.1 |
ENST00000580257.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010898.hg.1 |
NR_026901.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010898.hg.1 |
NR_026901.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008092.hg.1 |
ENST00000574657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000497090.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000497090.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
ENST00000497090.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000241014.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000395629.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000497090.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000497090.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
ENST00000497090.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007429.hg.1 |
NM_005456.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010898.hg.1 |
ENST00000580257.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010898.hg.1 |
ENST00000580257.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010898.hg.1 |
NR_026901.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010898.hg.1 |
NR_026901.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008092.hg.1 |
ENST00000574657.1 |
Ensembl |