| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006714.hg.1 |
ENST00000380272.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006714.hg.1 |
ENST00000380272.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006714.hg.1 |
ENST00000380272.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006714.hg.1 |
ENST00000380274.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006714.hg.1 |
ENST00000380274.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006714.hg.1 |
ENST00000380274.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006714.hg.1 |
ENST00000380274.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006714.hg.1 |
NM_012135.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006714.hg.1 |
NM_012135.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006714.hg.1 |
NM_012135.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006714.hg.1 |
ENST00000380272.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006714.hg.1 |
ENST00000380272.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006714.hg.1 |
ENST00000380272.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006714.hg.1 |
ENST00000380274.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006714.hg.1 |
ENST00000380274.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006714.hg.1 |
ENST00000380274.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006714.hg.1 |
ENST00000380274.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006714.hg.1 |
NM_012135.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006714.hg.1 |
NM_012135.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006714.hg.1 |
NM_012135.2 |
Refseq |